ContactUsLinkComponent
Thermometer, mercury, ASTM94C, white backed, measuring range: 80:110/0.1C, length:370mm, 76mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Catalog # 620-0988
Supplier:  Amarell
Thermometer, mercury, ASTM94C, white backed, measuring range: 80:110/0.1C, length:370mm, 76mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Catalog # 620-0988
Supplier:  Amarell