Thermometer, mercury, ASTM61C, white backed, measuring range: 32:127/0.2C, length:380mm, 79mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Thermometer, mercury, ASTM61C, white backed, measuring range: 32:127/0.2C, length:380mm, 79mm immersion, durable pigment, suitable for government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items