Lead, plasma standard solutions, Specpure®
Lead, Pb, 1000 mg/L, 5% HNO₃, Plastic bottle, 50 mL
Supplier: Thermo Fisher Scientific
Lead, plasma standard solutions, Specpure®
Catalog # 13853.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Pb in 5% HNO₃
UN No.: 3264
MDL No.: MFCD00134050
: Traceable to NIST
Specifications
Specifications
- Concentration:1000 mg/L
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Pk:50 mL
- Element:Lead
- Symbol:Pb
- Cat. No.:13853.AD
- MDL No.:MFCD00134050



