Orders LinkContactUsLinkComponent
Bottle Carriers
Bottle carrier, HDPE, Grey, Up to six bottles, 95 mm
  BURK0361-0006
 :   BURKLE
Bottle Carriers
  BURK0361-0006
 :  BURKLE
Bottle Carriers
Bottle Carriers
Bottle carrier, HDPE, Grey, Up to six bottles, 95 mm
15,20 €
1 * 1 items
Made of PP or HDPE.
  • Bottle transport container (PP) for up to 6 bottles, bottle transport box for up to 8 bottles
  • Bottle transport box (HDPE): suitable for food; With impermeable base suitable as a collecting basin
  • For:
    Up to six bottles
  • Colour:
    Grey
  • Description:
    Bottle carrier, HDPE
  • Container max. Ø:
    95 mm