Cap strips, domed top
Strips of 12 domed caps, Clear, 25 packs of 8
Catalog # 732-4899
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Suitable for use with 96-well PCR plates as well as standard and low profile tube strips.
- Made from Polypropylene (PP)
- Large end tabs for strip removal
- Certified free from DNase, RNase and human DNA
Documents
ADVAAB-0265_TDS_EN
Datasheet
Specifications
- Packed:25 packs of 8
- Colour:Clear
- Description:Strips of 12 domed caps
- Seq:06