Orders LinkContactUsLinkComponent
Spray bottles, LaboPlast®
Spray bottles, LaboPlast®
250 ml
Catalog # 215-6510
Supplier:   BURKLE
Spray bottles, LaboPlast®
Catalog # 215-6510
Supplier:  BURKLE
Spray bottles, LaboPlast®
Spray bottles, LaboPlast®
250 ml
Price
5,90 €
Log in for availability
1 * 1 items
Product Details & Documents
LDPE, translucent, PP spray attachment.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
  • Media-contacting parts PP, PE and stainless steel
Spray, choice of fine mist or focused jet spray functions to disperse fluids.
Specifications
  • Capacity:
    250 ml