Potassium hydroxide ca. 85%, pellets ACS
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Potassium hydroxide ca. 85%, pellets ACS
Catalog # ACRO424140025
Supplier: Thermo Fisher Scientific
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (760 mmHg)
- Melting Pt:360 °C
- Density:2,04 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1813
- ADR:8,II
- REACH:01-2119487136-33
Specifications
Specification Test Results
- Appearance:White Pellets
- Titration with HCl:≥85 % (KOH)
- Chloride (Cl):≤0.01 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.003 %
- Potassium carbonate:≤2.0 % (K2CO3)
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.002 %
- Nickel (Ni):≤0.001 %
- Sodium (Na):≤0.05 %
- Heavy metals (ICP-OES):≤0.001 %
- Nitrogen compounds (as N):≤0.001 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



