Molybdic acid ACS
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7782-91-4
- Formula:H₂MoO₄
- MDL Number:MFCD00036287
- Molecular Weight:161.95 g/mol
- Merck Index:13,06262
- Boiling Pt:~ 300 °C (1013 hPa)
- Melting Pt:300 °C
- Density:3.1 g/cm³
- Storage Temperature:Ambient
- EINECS:231-970-5
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration:≥85.0 % (Precipitation titraton of molybdate)
- Chloride (Cl):≤0.002 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.2 %
- AsO4, PO4, SiO4 (as SiO2):≤0.001 %
- Heavy metals (as Pb):≤0.003 %
- Insoluble matter:≤0.01 % (in dilute NH4OH)
Specifications
- Pack type:Plastic bottle
- Pk:500 g



