Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl red ACS

Methyl red ACS

Glass bottle for solids, 25 g
Catalog # ACRO423970250
Methyl red ACS
Catalog # ACRO423970250
CAS Number:  493-52-7
Methyl red ACS
Methyl red ACS

Glass bottle for solids, 25 g

CAS Number:  493-52-7
Price
42,50 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1,31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark red to brown to purple Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    179 to 182 °C
  • Visual transition interval:
    From pH 4.2 (pink) to pH 6.2 (yellow)
  • Clarity of alcohol solution:
    Passes test
  • Specific optical rotation:
    +52.0° to +62.0° (20 °C, 589 nm) (neat)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications