Lead(II) nitrate ACS
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Lead(II) nitrate ACS
Catalog # ACRO423855000
Supplier: Thermo Fisher Scientific
CAS Number: 10099-74-8
Jump to:
Product Details & Documents
Synonyms: Lead dinitrate
- CAS Number:10099-74-8
- Formula:Pb(NO₃)₂
- MDL Number:MFCD00011153
- Molecular Weight:331.21 g/mol
- Merck Index:13,05432
- Melting Pt:470 °C (dec.)
- Density:4.53 g/cm³
- Storage Temperature:Ambient
- EINECS:233-245-9
- UN:1469
- ADR:5.1,II
Documents
ACRO42385
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Complexometric:≥99.0 %
- Chloride (Cl):≤10 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤20 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤200 ppm
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



