Electroporation Cuvettes Plus
Cuvettes Plus with 1,0 mm gap size, grey cap, 20 - 90 µl, Sterile
732-0020
: BTX
High quality disposable cuvettes for transfection available in three sizes and compatible with electroporation systems.
- Transfer pipette for manipulation of samples and gamma irradiated together for sterility
- Polished aluminium electrodes in the cuvettes lowers arcing frequency
- Cuvettes (without cap) are autoclavable
Cuvette caps are round for easy, one-handed removal, and are colour-coded for quick identification.
- Sterility:Sterile
- Volume:20 - 90 µl
- Description:Cuvettes Plus with 1,0 mm gap size, grey cap
- Seq:1