Order Entry
Export
Orders LinkContactUsLinkComponent
Electrodes, Genetrodes™

Electrodes, Genetrodes™

Genetrode™, bent L-shaped, 1 mm*
Catalog # 732-0014
Supplier:   BTX
Electrodes, Genetrodes™
Catalog # 732-0014
Supplier:  BTX
Electrodes, Genetrodes™
Electrodes, Genetrodes™

Genetrode™, bent L-shaped, 1 mm*

Price
575,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Genetrodes™ are paired, reusable, needle style electrodes that are ideal for in vivo and in ovo electroporation applications, including drug and gene delivery.

  • Five models to suit the size and shape of the target electroporation area
  • Each model consists of a pair of electrodes configured as either straight or bent L-shaped electrodes with gold tips
  • The electrodes are placed into tissue following injection of the molecule of interest, and an electroporation pulse is delivered using an electroporation generator
  • The electric field introduced by the Genetrodes™ causes transient pores to form in the cells of the tissue, allowing uptake of the molecules into cells
  • Genetrodes™ are positioned in parallel at a predetermined gap in tissue using the Genetrode/Genepaddle holder

Generator compatibility: ECM® 830, ECM® 2001
Voltage range: 0 to 200 V DC

Pulse length range: 10 µsec to 99 msec in PBS
Pulse number range: 1 to 99 (depending on voltage and pulse length settings)

Genetrode™ holder electrode gap: 1 to 10 mm range.

* Requires Genetrodes/Genepaddle holder, square post cables and coaxial banana plug cable.

Genetrode™ holder electrode gap: 1 - 10 mm range

Documents

Specifications
  • Application:
    in ovo
  • Description:
    Genetrode™, bent L-shaped, 1 mm*
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Genetrode/Genepaddle holder
    Genetrode/Genepaddle holder
    732-0028
    Log in for availability
    1 * 1 items - 594,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Black and red electrode connection cable (Banana to Banana), 3 m, can be used with Genetrodes™, Genepaddles™ and flat electrode for cell fusion
    Black and red electrode connection cable (Banana to Banana), 3 m, can be used with Genetrodes™, Genepaddles™ and flat electrode for cell fusion
    BTXI45-0217
    Log in for availability
    1 * 1 items - 210,00 €