Orders LinkContactUsLinkComponent
Funnels for conical ground joints
Funnels for conical ground joints
19/26 NS, 50 mm
Catalog # 241-0232
Supplier:   Brand
Funnels for conical ground joints
Catalog # 241-0232
Supplier:  Brand
Funnels for conical ground joints
Funnels for conical ground joints
19/26 NS, 50 mm
Price
61,50 €
Log in for availability
1 * 10 items
Product Details & Documents
PP, translucent.
  • Ideal for transferring liquids or powders to multiple-neck flasks
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
Specifications
  • Funnel O-Ø flat side:
    50 mm
  • Stem for:
    19/26 NS