Order Entry
Export
Orders LinkContactUsLinkComponent
Thymol blue ACS

Thymol blue ACS

Glass bottle for solids, 5 g
Catalog # ACRO420930050
Thymol blue ACS
Catalog # ACRO420930050
CAS Number:  76-61-9
Thymol blue ACS
Thymol blue ACS

Glass bottle for solids, 5 g

CAS Number:  76-61-9
Price
37,20 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Melting Pt:221 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-973-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark green to dark brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Visual transition interval:
    from pH 1.2 (pink) to pH 2.8 (yellow)
  • Clarity of solution:
    passes test

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications