Micro pipettes, intraMARK, BLAUBRAND®
10 µl, 125 mm
Supplier: Brand
Jump to:
Product Details & Documents
Glass, with ring mark, disposable.
- Calibrated to contain 'In', contains the stated volume when filled from one end to a ring mark
- Colour coded to ISO for clear identification
- DE-M marking for volumes ≥5 µl
Accuracy ≤±0,25%; Reproducibility ≤0,5% (except for 612-1401: Accuracy ≤±0,30%; Reproducibility ≤0,6%).
DIN ISO 7550, IVD 98/79 EC
Caution: DE-M marking only for 5 µl.
Documents
Certificates
Search by Lot NumberBrand Volumetric Measurement in the Laboratory
Product Literature
BRND708757_U_TDS_EN
Datasheet
Specifications
- Capacity:10 µl
- Length:125 mm
- Seq:02



