Order Entry
Export
Orders LinkContactUsLinkComponent
 

Silver nitrate 99+% ACS

Glass bottle for solids, 100 g
  ACRO419351000
 
 
 
Silver nitrate 99+% ACS
  ACRO419351000
 :  7761-88-8
Silver nitrate 99+% ACS
Silver nitrate 99+% ACS

Glass bottle for solids, 100 g

 :  7761-88-8
 
392,00 €
   
1 * 100 g
 
 :  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  •  :7761-88-8
  •  :AgNO₃
  •  :MFCD00003414
  •  :169.87 g/mol
  •  :12,08661
  •  :444 °C (760 mmHg)
  •  :212 °C
  •  :4,35 g/cm³ (20 °C)
  •  :Ambient
  •  :231-853-9
  •  :1493
  •  :5.1,II
  •  :01-2119513705-43

 

 

 

  • Appearance:
    White Crystalline powder or crystals
  • Free acid:
    Passes test
  • Titration:
    ≥99.0 %
  • Clarity of solution:
    Passes test
  • Chloride (Cl):
    ≤5 ppm
  • Sulfate (SO4):
    ≤20 ppm
  • Copper (Cu):
    ≤2 ppm
  • Iron (Fe):
    ≤2 ppm
  • Lead (Pb):
    ≤10 ppm
  • Subst not pptd by HCl:
    ≤0.01 %

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g