Orders LinkContactUsLinkComponent
Repair set
Repair set
Repair set for Transferpettor™ 100, 200 µl
Catalog # 612-5385
Supplier:   Brand
Repair set
Catalog # 612-5385
Supplier:  Brand
Repair set
Repair set
Repair set for Transferpettor™ 100, 200 µl
Price
36,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Set includes 1 allen key, 1 piston rod with fitted PTFE Transferpettor™ seal (for capacities ≥ 20 µl), 1 calibration gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE Transferpettor™ seals, 1 mounting block (for capacities ≥ 20 µl).
Documents
Specifications
  • Description:
    Repair set for Transferpettor™ 100, 200 µl
  • Seq:
    05