Phenol red ACS
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:205-609-7
Documents
Specifications
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 6.8 (yellow) to pH 8.2 (red)
- Clarity of solution:Passes test
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



