Orders LinkContactUsLinkComponent
Transferpettor™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
Transferpettor™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
20 - 100 µl, 0,1 µl, ±0,6 %, ≤0,4 %, Blue, 200 µl
Catalog # 613-3357
Supplier:   Brand
Transferpettor™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
Catalog # 613-3357
Supplier:  Brand
Transferpettor™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
Transferpettor™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
20 - 100 µl, 0,1 µl, ±0,6 %, ≤0,4 %, Blue, 200 µl
Price
382,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Ideal for pipetting highly viscous media up to 140000 mm²/s, or with density up to 13,6 g/cm³ (protein solutions, oils, fat, mercury, sulphuric acid), high vapour pressure media up to 500 mbar (alcohols, ethers, hydrocarbons), or media with a tendency to foam (tensioactive solutions).
  • For pipetting media that present problems for air displacement pipettes
  • Positive displacement ensures that results are reproducible regardless of pipetting speed and ambient conditions
  • The movable piston is in direct contact with the liquid and removes the last drop from the walls of the capillaries or tips
  • Residual moisture in the capillaries or tips is minimal and may be disregarded except in the case of infectious or radioactive media
  • DE-M marking, with performance certificate
Ordering information: Must be used with capillaries, tips and pistons for Transferpettor™.
Specifications
  • Colour code:
    Blue
  • Volume:
    20 - 100 µl
  • Seq:
    4
  • Increment:
    0,1 µl
  • Accuracy:
    ±0,6 %
  • Imprecision:
    ≤0,4 %
  • Recommended tips:
    200 µl
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PTFE, with mounting block, for 100/200 µl
    Transferpettor™ seals, PTFE, with mounting block, for 100/200 µl
    613-3377
    Log in for availability
    1 * 3 items - 18,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PE, for 100 - 500 µl
    Transferpettor™ seals, PE, for 100 - 500 µl
    613-3387
    Log in for availability
    1 * 10 items - 22,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PE, for 200 - 1000 µl
    Transferpettor™ seals, PE, for 200 - 1000 µl
    613-3388
    Log in for availability
    1 * 10 items - 22,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PE, for 1000 - 5000 µl
    Transferpettor™ seals, PE, for 1000 - 5000 µl
    613-3389
    Log in for availability
    1 * 10 items - 22,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PE, for 2000 - 10000 µl
    Transferpettor™ seals, PE, for 2000 - 10000 µl
    613-3390
    Log in for availability
    1 * 10 items - 22,60 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3370
    10 µl
    Orange
    Log in for availability
    1 * 100 items - 35,60 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3369
    1 / 2 / 5 µl
    White
    Log in for availability
    1 * 100 items - 35,60 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3371
    20 µl
    Black
    Log in for availability
    1 * 100 items - 35,60 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3373
    50 µl
    Green
    Log in for availability
    1 * 100 items - 35,60 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3374
    100 / 200 µl
    Blue/red
    Log in for availability
    1 * 50 items - 17,80 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • PP caps
    PP caps
    613-3383
    100 - 500 µl
    Green
    Log in for availability
    1 * 10 items - 29,40 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • PP caps
    PP caps
    613-3386
    2000 - 10000 µl
    Orange
    Log in for availability
    1 * 10 items - 29,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PTFE, with mounting block, for 20/25 µl
    Transferpettor™ seals, PTFE, with mounting block, for 20/25 µl
    613-3375
    Log in for availability
    1 * 3 items - 18,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Transferpettor™ seals, PTFE, with mounting block, for 50 µl
    Transferpettor™ seals, PTFE, with mounting block, for 50 µl
    613-3376
    Log in for availability
    1 * 3 items - 18,10 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • PP caps
    PP caps
    613-3385
    1000 - 5000 µl
    Red
    Log in for availability
    1 * 10 items - 29,40 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • PP caps
    PP caps
    613-3384
    200 - 1000 µl
    Yellow
    Log in for availability
    1 * 10 items - 29,40 €
Product Information
  • Catalog #
    Capacity
    Colour code
    Availability
    Price
  • Glass capillaries
    Glass capillaries
    613-3372
    25 µl
    2 × white
    Log in for availability
    1 * 100 items - 35,60 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • 613-3378
    4× Transferpettor™, up to 200 µl, with accessories
    Log in for availability
    1 * 1 items - 225,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • 613-3391
    2× Transferpettor™, 0,5 to 10 ml, with accessories
    Log in for availability
    1 * 1 items - 148,00 €