Pectin pract.
Plastic bottle, 1 kg
CAS No.: 9000-69-5
- :9000-69-5
- :MFCD00081838
- :Ambient
- Appearance:Off-white to beige to golden Powder
- Assay:67 to 71 % (degree of esterification)
- Loss on drying:≤12 % (105°C, 2 hrs)
- Particle size:≤2 % ( 60 mesh)
- Viscosity:30.5 to 36.5
- pH:3.0 to 3.8 (1 % solution)
- Pack type:Plastic bottle
- Pk:1 kg



