Paraformaldehyde 96%, extra pure
Plastic bottle, 25 g
: Polyoxymethylene
- :30525-89-4
- :(CH₂O)n
- :MFCD00133991
- :13,07096
- :135 - 136 °C (13 -rr)
- :120…170 °C (dec.)
- :1,46 g/cm³ (20 °C)
- :71 °C
- :Ambient
- :2213
- :4.1,III
- Appearance:White Powder or beads
- Infrared spectrum:Conforms
- Ash:≤0.02 %
- pH:4.0 to 5.5 (10 % suspension)
- Titration with HCl:≥95 %
- Free acid (HCOOH):≤0.03 %
- Pack type:Plastic bottle
- Pk:25 g



