Paraformaldehyde 96%, extra pure
Plastic bottle, 3 kg
Supplier: Thermo Fisher Scientific
Paraformaldehyde 96%, extra pure
Catalog # ACRO416780030
Supplier: Thermo Fisher Scientific
CAS Number: 30525-89-4
Jump to:
Product Details & Documents
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:13,07096
- Boiling Pt:135 - 136 °C (13 -rr)
- Melting Pt:120…170 °C (dec.)
- Density:1,46 g/cm³ (20 °C)
- Flash Pt:71 °C
- Storage Temperature:Ambient
- UN:2213
- ADR:4.1,III
Documents
Specifications
Specification Test Results
- Appearance:White Powder or beads
- Infrared spectrum:Conforms
- Ash:≤0.02 %
- pH:4.0 to 5.5 (10 % suspension)
- Titration with HCl:≥95 %
- Free acid (HCOOH):≤0.03 %
Specifications
- Pack type:Plastic bottle
- Pk:3 kg



