Desiccators, with lid
Dessicator, with PC lid, PP base and stopcock, 200 DN, 235×240 mm
Catalog # BRND65810
Supplier: Brand
Jump to:
Product Details & Documents
Dessicators with polycarbonate lid, PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobutadiene rubber (CK) O-ring, compressed by vacuum.
- Light-weight and easy to handle
Specifications
- Nominal size:200 DN
- Type:Dessicator, with PC lid, PP base and stopcock
- Ø ext.×H:235×240 mm
Accessories
Product Information
- Catalog #TypeForAvailabilityPrice
- Venting stopperVenting stopperBRND65804Venting stopper, PCDesiccatorsLog in for availability1 * 1 items - 21,90 €
Product Information
- Catalog #TypeØAvailabilityPrice
- Sealing ringSealing ringBRND65820Sealing ring, polychlorobutadiene rubber150 mmLog in for availability1 * 1 items - 12,90 €
Product Information
- Catalog #TypeØAvailabilityPrice
- Sealing ringSealing ringBRND65824Sealing ring, polychlorobutadiene rubber250 mmLog in for availability1 * 1 items - 19,60 €
Product Information
- Catalog #TypeØAvailabilityPrice
- Sealing ringSealing ringBRND65822Sealing ring, polychlorobutadiene rubber200 mmLog in for availability1 * 1 items - 16,90 €