Order Entry
Export
Orders LinkContactUsLinkComponent
Neutral red, pure, certified

Neutral red, pure, certified

Glass bottle for solids, 25 g
Catalog # ACRO415490250
Neutral red, pure, certified
Catalog # ACRO415490250
CAS Number:  553-24-2
Neutral red, pure, certified
Neutral red, pure, certified

Glass bottle for solids, 25 g

CAS Number:  553-24-2
Price
83,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:12,06571
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:209-035-8

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥50 %
  • UV Lambda max:
    539 to 544 nm
  • Absorption Ratio P-15/P+15:
    1 to 1.12
  • BSC Certification:
    Passes

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications