Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl oleate, technical

Methyl oleate, technical

Glass bottle for solids, 1 kg
Catalog # ACRO414640010
Methyl oleate, technical
Catalog # ACRO414640010
CAS Number:  67762-38-3
Methyl oleate, technical
Methyl oleate, technical

Glass bottle for solids, 1 kg

CAS Number:  67762-38-3
Price
59,80 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
  • CAS Number:67762-38-3
  • Formula:C₁₉H₃₆O₂
  • MDL Number:MFCD00009578
  • Molecular Weight:296.49 g/mol
  • Merck Index:12,06965
  • Boiling Pt:186 °C (9 mmHg)
  • Melting Pt:–20 °C
  • Density:0.87 g/cm³
  • Flash Pt:170 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-992-5

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to amber Viscous liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥55 %
  • HPLC:
    ≥98.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications