Order Entry
Export
Orders LinkContactUsLinkComponent
MBD 98%

MBD 98%

Glass bottle, 1 g
Catalog # ACRO413540010
MBD 98%
Catalog # ACRO413540010
CAS Number:  33984-50-8
MBD 98%
MBD 98%

Glass bottle, 1 g

CAS Number:  33984-50-8
Price
122,00 €
Log in for availability
1 * 1 g
Product Details & Documents

CAS No.: 33984-50-8

Synonyms:  7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole
  • CAS Number:33984-50-8
  • Formula:C₁₄H₁₂N₄O₄
  • MDL Number:MFCD00042778
  • Molecular Weight:300.274 g/mol
  • Melting Pt:176…180 °C
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    Orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    177 to 182 °C
  • HPLC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications