Order Entry
Export
Orders LinkContactUsLinkComponent
Linoleic acid 60% tech.

Linoleic acid 60% tech.

Glass bottle for solids, 500 g
Catalog # ACRO413195000
Linoleic acid 60% tech.
Catalog # ACRO413195000
CAS Number:  60-33-3
Linoleic acid 60% tech.
Linoleic acid 60% tech.

Glass bottle for solids, 500 g

CAS Number:  60-33-3
Price
65,50 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS Number:60-33-3
  • Formula:C₁₈H₃₂O₂
  • MDL Number:MFCD00064241
  • Molecular Weight:280.45 g/mol
  • Boiling Pt:229…230 °C (16 mmHg)
  • Melting Pt:–5 °C
  • Density:0.9 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-470-9

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≤3 % C18 (sat.)
  • Acid value:
    195 to 205 mg KOH/g
  • Iodine value:
    130 to 148 g I2/100 g

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications