Calcein
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
- CAS Number:1461-15-0
- Formula:C₃₀H₂₆N₂O₁₃
- MDL Number:MFCD00005049
- Molecular Weight:622.53 g/mol
- Melting Pt:200 °C
- Storage Temperature:Ambient
- EINECS:215-957-1
Documents
Specifications
Specification Test Results
- Appearance:Yellow to orange-brown Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Suitability:(for Ca determination) Passes test
Specifications
- Pack type:Glass bottle
- Pk:25 g



