Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl ether 99+% stabilised for spectroscopy

Diethyl ether 99+% stabilised for spectroscopy

Glass bottle for solids, 1 kg
Catalog # ACRO410050010
Irritant
Flammable
Diethyl ether 99+% stabilised for spectroscopy
Catalog # ACRO410050010
CAS Number:  60-29-7
Diethyl ether 99+% stabilised for spectroscopy
Diethyl ether 99+% stabilised for spectroscopy

Glass bottle for solids, 1 kg

CAS Number:  60-29-7
Price
83,70 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:34.6 °C (760 mmHg)
  • Melting Pt:–116 °C
  • Density:0.714 g/cm³
  • Flash Pt:–45 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with ethanol
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • GC:
    ≥99.0 % (Excluding stabilizer)
  • UV:
    at 220 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    Approx. 1 % w/w Ethanol
  • Peroxides:
    ≤5 ppm (as H2O2)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications