Order Entry
Export
Orders LinkContactUsLinkComponent
Bulb pipettes, BLAUBRAND®, two marks, class AS

Bulb pipettes, BLAUBRAND®, two marks, class AS

20 ml, 0,03 ± ml, 510 mm, Yellow
Catalog # 612-2732
Supplier:   Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Catalog # 612-2732
Supplier:  Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Bulb pipettes, BLAUBRAND®, two marks, class AS

20 ml, 0,03 ± ml, 510 mm, Yellow

Price
14,10 €
Log in for availability
1 * 1 items
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate

DIN EN ISO 648

Certifications: DIN EN ISO 648; DE-M marking.

Specifications
  • Colour code:
    Yellow
  • Capacity:
    20 ml
  • Length:
    510 mm
  • Seq:
    08
  • Tolerance:
    0,03 ± ml