Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bulb pipettes, BLAUBRAND®, two marks, class AS

2 ml, 0,01 ± ml, 330 mm, Orange
Catalog # 612-2728
Supplier:   Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Catalog # 612-2728
Supplier:  Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Bulb pipettes, BLAUBRAND®, two marks, class AS

2 ml, 0,01 ± ml, 330 mm, Orange

Price
8,60 €
Log in for availability
1 * 1 items
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate

DIN EN ISO 648

Certifications: DIN EN ISO 648; DE-M marking.

Specifications
  • Colour code:
    Orange
  • Capacity:
    2 ml
  • Length:
    330 mm
  • Seq:
    03
  • Tolerance:
    0,01 ± ml