Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

69 mm, 10 mm, 80 mm
Catalog # BRND148225
Supplier:   Brand
Funnels with short stem
Catalog # BRND148225
Supplier:  Brand
Funnels with short stem
Funnels with short stem

69 mm, 10 mm, 80 mm

Price
140,00 €
Log in for availability
1 * 20 items
Product Details & Documents

PP, translucent.

  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers

For rapid filtration.

Specifications
  • Funnel O-Ø:
    80 mm
  • Stem O-Ø:
    10 mm
  • Stem length:
    69 mm