Order Entry
Export
Orders LinkContactUsLinkComponent
 

Funnels with short stem

95 mm, 17 mm, 150 mm
Catalog # BRND147035
Supplier:   Brand
Funnels with short stem
Catalog # BRND147035
Supplier:  Brand
Funnels with short stem
Funnels with short stem

95 mm, 17 mm, 150 mm

Price
48,60 €
Log in for availability
1 * 12 items
Product Details & Documents

PP, translucent.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

With smooth interior surfaces.

Specifications
  • Funnel O-Ø:
    150 mm
  • Stem O-Ø:
    17 mm
  • Stem length:
    95 mm