Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

77 mm, 13 mm, 100 mm
Catalog # BRND147025
Supplier:   Brand
Funnels with short stem
Catalog # BRND147025
Supplier:  Brand
Funnels with short stem
Funnels with short stem

77 mm, 13 mm, 100 mm

Price
26,40 €
Log in for availability
1 * 12 items
Product Details & Documents

PP, translucent.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

With smooth interior surfaces.

Specifications
  • Funnel O-Ø:
    100 mm
  • Stem O-Ø:
    13 mm
  • Stem length:
    77 mm