Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

55 mm, 10 mm, 75 mm
Catalog # BRND147020
Supplier:   Brand
Funnels with short stem
Catalog # BRND147020
Supplier:  Brand
Funnels with short stem
Funnels with short stem

55 mm, 10 mm, 75 mm

Price
16,20 €
Log in for availability
1 * 12 items
Product Details & Documents

PP, translucent.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

With smooth interior surfaces.

Specifications
  • Funnel O-Ø:
    75 mm
  • Stem O-Ø:
    10 mm
  • Stem length:
    55 mm