Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

43 mm, 9 mm, 50 mm
Catalog # BRND147015
Supplier:   Brand
Funnels with short stem
Catalog # BRND147015
Supplier:  Brand
Funnels with short stem
Funnels with short stem

43 mm, 9 mm, 50 mm

Price
28,80 €
Log in for availability
1 * 24 items
Product Details & Documents

PP, translucent.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

With smooth interior surfaces.

Specifications
  • Funnel O-Ø:
    50 mm
  • Stem O-Ø:
    9 mm
  • Stem length:
    43 mm