Stoppers for standard ground joints
24/29
Catalog # 217-9268
Supplier: Brand
Jump to:
Product Details & Documents
PP or PE-LD, hollow, with sealing ribs and square or octagonal grip.
- Good chemical resistance, PP stoppers are autoclavable (+121 °C)
- Sealing rings for tight, leakproof fit
- Square grip (up to sizes 29/32, PP) or octagonal grip (sizes 34/35 and larger, PE-LD) for easy release and removal of the stopper
Specifications
- NS:24/29