Charcoal activated, NORIT® SA 2, decolorizing
Glass bottle for solids, 500 g
: Carbon activated, Activated carbon, Activated charcoal
- :7440-44-0
- :C
- :MFCD00133992
- :12.01 g/mol
- :13,01818
- :3652 °C
- :2 g/cm³ (20 °C)
- :Ambient
- :231-153-3
- :4.2,III
- :01-2119488894-16
Documents
ACRO40403
SDS
- Appearance:Black Powder
- Loss on drying:≤10 %
- Molasses number:≤390
- GC:≥95 %
- Pack type:Glass bottle for solids
- Pk:500 g



