Charcoal activated, DARCO® 4-12 mesh, decolorizing
Plastic drum, 3 kg
Supplier: Thermo Fisher Scientific
Charcoal activated, DARCO® 4-12 mesh, decolorizing
Catalog # ACRO403990030
Supplier: Thermo Fisher Scientific
CAS Number: 7440-44-0
Jump to:
Product Details & Documents
Synonyms: Carbon activated, Activated carbon, Activated charcoal
- CAS Number:7440-44-0
- Formula:C
- MDL Number:MFCD00133992
- Molecular Weight:12.01 g/mol
- Merck Index:13,01818
- Melting Pt:3652 °C
- Density:2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-153-3
- ADR:4.2,III
- REACH:01-2119488894-16
Documents
ACRO40399
SDS
Specifications
Specification Test Results
- Appearance:Black Granules and/or chunks
- Loss on drying:≤12 %
- Particle size:≤5 % (> 4 mesh)
- pH:3.5 to 6
Specifications
- Pack type:Plastic drum
- Pk:3 kg



