Storage and Dispenser Boxes
Small, 165×152×178 mm
216-2848
: Brand
PMMA, white sides, transparent front.
- Easy top loading and convenient dispensing through the front flap
- Clear front allows contents to be checked easily
- Cover and front flap limit contamination by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.
- Description:Small
- W×D×H int.:165×152×178 mm