Orders LinkContactUsLinkComponent
Storage and Dispenser Boxes
Storage and Dispenser Boxes
Large, 165×152×355 mm
Catalog # 216-2847
Supplier:   Brand
Storage and Dispenser Boxes
Catalog # 216-2847
Supplier:  Brand
Storage and Dispenser Boxes
Storage and Dispenser Boxes
Large, 165×152×355 mm
Price
258,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PMMA, white sides, transparent front.
  • Easy top loading and convenient dispensing through the front flap
  • Clear front allows contents to be checked easily
  • Cover and front flap limit contamination by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.
Specifications
  • Description:
    Large
  • W×D×H int.:
    165×152×355 mm