Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Burette, BLAUBRAND, class AS, DE-M marking, straight stopcock, 50 ml:0,1 ml, Boro 3.3, Schellbach stripes, glass key 1 * 1 items
612-9963
:
Brand
Burette, BLAUBRAND, class AS, DE-M marking, straight stopcock, 50 ml:0,1 ml, Boro 3.3, Schellbach stripes, glass key 1 * 1 items
612-9963
:
Brand
BLAUBRAND® Volumetric Instruments
Brand Volumetric Measurement in the Laboratory