(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Catalog # ACRO344580010
Supplier: Thermo Fisher Scientific
CAS Number: 7326-19-4
Jump to:
Product Details & Documents
Synonyms: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-(+)-Phenyllactic acid
- CAS Number:7326-19-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00078062
- Molecular Weight:166.1739 g/mol
- Melting Pt:121…125 °C
- Storage Temperature:Ambient
- EINECS:230-803-3
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:121 to 125 °C
- Specific optical rotation:+17° to +20° (20 °C, 589 nm) (c=1, CH3OH)
- Titration with NaOH:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



