Benzophenone oxime 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:574-66-3
- Formula:C₁₃H₁₁NO
- MDL Number:MFCD00051461
- Molecular Weight:197.24 g/mol
- Melting Pt:140…144 °C
- Storage Temperature:Freezer
- EINECS:209-373-6
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:140 to 144 °C
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



