4-Hydroxy-2-methoxybenzaldehyde 98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4-Hydroxy-2-methoxybenzaldehyde 98%
Catalog # ACRO335930050
Supplier: Thermo Fisher Scientific
CAS Number: 18278-34-7
Jump to:
Product Details & Documents
- CAS Number:18278-34-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00051964
- Molecular Weight:152.15 g/mol
- Boiling Pt:273 °C (1013 hPa)
- Melting Pt:153…158 °C
- Storage Temperature:Ambient
Documents
ACRO33593
SDS
Specifications
Specification Test Results
- Appearance:Off-white to light brown Powder
- Infrared spectrum:Conforms
- Melting point:156 to 161 °C
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



