Diphenylsilane 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:775-12-2
- Formula:C₁₂H₁₂Si
- MDL Number:MFCD00003002
- Molecular Weight:184.31 g/mol
- Boiling Pt:95…97 °C (13 mmHg)
- Density:0.99 g/cm³
- Flash Pt:98 °C
- Storage Temperature:Ambient
- EINECS:212-271-4
Documents
ACRO32977
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5780 to 1.5800 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



