Acetone for residue analysis
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Acetone for residue analysis
Catalog # ACRO326570010
Supplier: Thermo Fisher Scientific
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.2 %
- GC:(ECD) Passes test, (NPD) Passes test, (FID) Passes test
- Residue after evaporation:≤2 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ACRO326570010
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



