PTFE balls, BOLA
Balls, PTFE, 6 mm
Catalog # BOHLH964-06
Supplier: Bohlender
Jump to:
Product Details & Documents
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
Specifications
- Ø:6 mm
- Description:Balls
- Material:PTFE
- Seq:004