Orders LinkContactUsLinkComponent
Laboratory trays
Laboratory trays
450×320×80 mm, 400×270×80 mm
Catalog # 216-0049
Supplier:   BOCHEM
Laboratory trays
Catalog # 216-0049
Supplier:  BOCHEM
Laboratory trays
Laboratory trays
450×320×80 mm, 400×270×80 mm
Price
130,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • Rounded corners for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, excellent thermal resistance
Multipurpose trays for a wide range of uses. Ideal as developing trays.
Specifications
  • W×D×H ext.:
    450×320×80 mm
  • Seq:
    06
  • W×D×H int.:
    400×270×80 mm