Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 6,5 l pots
Catalog # BOCH8373
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10.
- Rounded corners for fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
: Lids to be ordered separately.
Specifications
- Description:Lids for laboratory pots, stainless steel 18/10, for 6,5 l pots
- Seq:12